Hesperidin

Lieferant: Roth Carl

ROTH7577.1EA 201 EUR
ROTH7577.1
Hesperidin
Hesperidin
Formel: C₂₈H₃₄O₁₅
Molekulargewicht: 610,56 g/mol
MDL Number: MFCD00075663
CAS-Nummer: 520-26-3
EINECS: 208-288-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR