N(α)-Cbz-L-histidin

Lieferant: Molekula

MOLE10028802-5GEA 0 EUR
MOLE10028802-5G
N(α)-Cbz-L-histidin
N(α)-Cbz-L-histidin
Formel: C₁₄H₁₅N₃O₄
Molekulargewicht: 289,3 g/mol
MDL Number: MFCD00065960
CAS-Nummer: 14997-58-1
EINECS: 239-084-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR