(Triphenylmethyl)triphenylphosphoniumchlorid ≥98%
|
Achtung
|
B22319.30EA
656
EUR
B22319.30
B22319.09
B22319.18
(Triphenylmethyl)triphenylphosphoniumchlorid ≥98%
(Triphenylmethyl)triphenylphosphoniumchlorid
Formel:
C₃₇H₃₀ClP Schmelzpunkt: 244…245 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00082244 CAS-Nummer: 48234-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...