7-(2-Hydroxyethyl)theophyllin

Lieferant: Molekula

MOLEM85896596EA 0 EUR
MOLEM85896596
7-(2-Hydroxyethyl)theophyllin
7-(2-Hydroxyethyl)theophyllin
Formel: C₉H₁₂N₄O₃
MDL Number: MFCD00055055
CAS-Nummer: 519-37-9
EINECS: 208-269-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR