Chlorfenapyr, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

37913-100MG-REA 77.2 EUR
37913-100MG-R
Chlorfenapyr, Supelco®
Chlorfenapyr
Formel: C₁₅H₁₁BrClF₃N₂O
MDL Number: MFCD01631152
CAS-Nummer: 122453-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR