2-Hydroxy-5-methylbenzophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIALH38608-5GEA
75
EUR
SIALH38608-5G
2-Hydroxy-5-methylbenzophenon, Sigma-Aldrich®
2-Hydroxy-5-methylbenzophenon
Formel:
C₁₄H₁₂O₂ |
MDL Number:
MFCD00002379 CAS-Nummer: 1470-57-1 EINECS: 216-002-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...