3,4-Dimethylbenzophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIALD149675-50GEA
161
EUR
SIALD149675-50G
3,4-Dimethylbenzophenon, Sigma-Aldrich®
3,4-Dimethylbenzophenon
Formel:
C₁₅H₁₄O Molekulargewicht: 210,28 g/mol |
MDL Number:
MFCD00008525 CAS-Nummer: 2571-39-3 EINECS: 219-916-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...