(S)-(+)-2,2-Dimethylcyclopropancarbonsäureamid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL434639-10GEA
90
EUR
SIAL434639-10G
(S)-(+)-2,2-Dimethylcyclopropancarbonsäureamid, Sigma-Aldrich®
(S)-(+)-2,2-Dimethylcyclopropancarbonsäureamid
Formel:
C₆H₁₁NO Molekulargewicht: 113,1600 g/mol |
MDL Number:
MFCD00216614 CAS-Nummer: 75885-58-4 EINECS: 278-334-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...