Allantoin

Lieferant: U.S. Pharmacopeia

USPH1012939EA 353 EUR
USPH1012939
Allantoin
Allantoin
Formel: C₄H₆N₄O₃
Molekulargewicht: 158,12
MDL Number: MFCD00005260
CAS-Nummer: 97-59-6
EINECS: 202-592-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR