10,12-Tricosadiinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 10,12-Tricosadiynoicacid

91445-5G
SIAL91445-5GEA 186 EUR
SIAL91445-5G
10,12-Tricosadiinsäure, Sigma-Aldrich®
10,12-Tricosadiynoic acid
Formel: C₂₃H₃₈O₂
Molekulargewicht: 346,56 g/mol
MDL Number: MFCD00041683
CAS-Nummer: 66990-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR