Guanin Hydrochlorid

Lieferant: Roth Carl

ROTH3814.1EA 27.3 EUR
ROTH3814.1
Guanin Hydrochlorid
Guanin Hydrochlorid
Formel: C₅H₆ClN₅O
Molekulargewicht: 187,59 g/mol
MDL Number: MFCD00213670
CAS-Nummer: 635-39-2
EINECS: 211-235-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR