3-Iod-1-methyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

SIAL729396-500MGEA 133 EUR
SIAL729396-500MG
3-Iod-1-methyl-1H-pyrazol, Sigma-Aldrich®
3-Iodo-1-methyl-1H-pyrazole
Formel: C₄H₅IN₂
Molekulargewicht: 208,00 g/mol
MDL Number: MFCD08435890
CAS-Nummer: 92525-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR