2-Amino-5-pyrimidinboronsäure

Lieferant: Thermo Scientific

MAYBMAY00047DAEA 0 EUR
MAYBMAY00047DA
2-Amino-5-pyrimidinboronsäure
2-Amino-5-pyrimidineboronic acid
Formel: C₄H₆BN₃O₂
Molekulargewicht: 138,92
MDL Number: MFCD07375147
CAS-Nummer: 936250-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR