4-Isopropyl-5-pyrimidinboronsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
H53118.MDEA
381
EUR
H53118.MD
4-Isopropyl-5-pyrimidinboronsäure 95%
4-Isopropyl-5-pyrimidinboronsäure
Formel:
C₇H₁₁BN₂O₂ Molekulargewicht: 165,99 Schmelzpunkt: 104…109 °C |
MDL Number:
MFCD08235084 CAS-Nummer: 913835-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...