1-Naphthylacetamid

Lieferant: Molekula

MOLEM33249429EA 0 EUR
MOLEM33249429
1-Naphthylacetamid
1-Naphthylacetamid
Formel: C₁₂H₁₁NO
Molekulargewicht: 185,23 g/mol
MDL Number: MFCD00004047
CAS-Nummer: 86-86-2
EINECS: 201-704-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR