Tris[N,N-bis(trimethylsilyl)amid]yttrium(III), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL524514-5GEA
476
EUR
SIAL524514-5G
Tris[N,N-bis(trimethylsilyl)amid]yttrium(III), Sigma-Aldrich®
Tris[N,N-bis(trimethylsilyl)amid]yttrium(III)
Formel:
C₁₈H₅₄N₃Si₆Y Molekulargewicht: 570,06 g/mol |
MDL Number:
MFCD00210649 CAS-Nummer: 41836-28-6 UN-Nummer: 3396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...