4-(N,N-Diphenylamino)benzaldehyd, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL647209-5GEA
150
EUR
SIAL647209-5G
4-(N,N-Diphenylamino)benzaldehyd, Sigma-Aldrich®
4-(N,N-Diphenylamino)benzaldehyd
Formel:
C₁₉H₁₅NO Molekulargewicht: 273,33 g/mol |
MDL Number:
MFCD00145131 CAS-Nummer: 4181-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...