2-Fluor-5-formylphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

SIAL715328-500MGEA 94.8 EUR
SIAL715328-500MG
2-Fluor-5-formylphenylboronsäurepinakolester, Sigma-Aldrich®
2-Fluor-5-formylphenylboronsäurepinakolester
Formel: C₁₃H₁₆BFO₃
Molekulargewicht: 250,07 g/mol
MDL Number: MFCD11867877
CAS-Nummer: 1112208-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR