3-Fluor-2-formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL709832-1GEA 64.4 EUR
SIAL709832-1G
3-Fluor-2-formylphenylboronsäure, Sigma-Aldrich®
3-Fluor-2-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molekulargewicht: 167,93 g/mol
MDL Number: MFCD10697421
CAS-Nummer: 871126-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR