Tolylfluanid
Lieferant: EHRENSTORFER GMBH
EHERC17600000EA
95.5
EUR
EHERC17600000
Tolylfluanid
Tolylfluanid
Formel:
C₁₀H₁₃Cl₂FN₂O₂S₂ |
MDL Number:
MFCD00055477 CAS-Nummer: 731-27-1 EINECS: 211-986-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...