Tolylfluanid, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

32060-250MGEA 82 EUR
32060-250MG
Tolylfluanid, Supelco®
Tolylfluanid
Formel: C₁₀H₁₃Cl₂FN₂O₂S₂
MDL Number: MFCD00055477
CAS-Nummer: 731-27-1
EINECS: 211-986-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR