2-Amino-4'-methoxyacetophenon Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL415944-1GEA 35.8 EUR
SIAL415944-1G SIAL415944-5G
2-Amino-4'-methoxyacetophenon Hydrochlorid, Sigma-Aldrich®
2-Amino-4'-methoxyacetophenon Hydrochlorid
Formel: C₉H₁₂ClNO₂
Molekulargewicht: 201,65 g/mol
MDL Number: MFCD00193078
CAS-Nummer: 3883-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR