2',3'-Dihydroxy-4'-methoxyacetophenon Hydrat, Sigma-Aldrich®

Lieferant: Merck

SIAL711225-25GEA 245 EUR
SIAL711225-25G
2',3'-Dihydroxy-4'-methoxyacetophenon Hydrat, Sigma-Aldrich®
2',3'-Dihydroxy-4'-methoxyacetophenon Hydrat
Formel: C₉H₁₀O₄·H₂O
MDL Number: MFCD00180772
CAS-Nummer: 708-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR