4'-Hydroxy-3'-iodacetophenon, Sigma-Aldrich®

Lieferant: Merck

SIAL722057-5GEA 127 EUR
SIAL722057-5G
4'-Hydroxy-3'-iodacetophenon, Sigma-Aldrich®
4'-Hydroxy-3'-iodacetophenon
Formel: C₉H₉IO₂
MDL Number: MFCD06200221
CAS-Nummer: 62615-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR