2-Aminoazotoluol 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Solvent Yellow 3
, 4′-Amino-2,3′-dimethylazobenzene
, Fast Garnet GBC base
, C.I. 11160
ACRO153260250EA
40.8
EUR
ACRO153260250
2-Aminoazotoluol 97%
2-Aminoazotoluol
Formel:
C₁₄H₁₅N₃ Molekulargewicht: 225,29 g/mol Schmelzpunkt: 101…102 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007733 CAS-Nummer: 97-56-3 EINECS: 202-591-2 |
Spezifikation Testergebnisse
Appearance | Orange to red-brown to dark brown Powder |
Infrared spectrum | Conforms |
Melting point | 100 to 104 °C |
Loss on drying | ≤3.0 % (110 °C, 1 h) |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...