2-Amino-4'-brombenzophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL676063-5GEA 127 EUR
SIAL676063-5G
2-Amino-4'-brombenzophenon, Sigma-Aldrich®
2-Amino-4'-brombenzophenon
Formel: C₁₃H₁₀BrNO
Molekulargewicht: 276,1300 g/mol
MDL Number: MFCD02181034
CAS-Nummer: 1140-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR