3-Brom-4'-fluorbenzophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL550493-1GEA 95.7 EUR
SIAL550493-1G
3-Brom-4'-fluorbenzophenon, Sigma-Aldrich®
3-Brom-4'-fluorbenzophenon
Formel: C₁₃H₈BrFO
MDL Number: MFCD00672034
CAS-Nummer: 46698-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR