5-Chlor-2-(methylamino)benzophenon
Lieferant: LGC Standards PROMOCHEM
PROOBP378EA
0
EUR
PROOBP378
5-Chlor-2-(methylamino)benzophenon
5-Chlor-2-(methylamino)benzophenon
Formel:
C₁₄H₁₂ClNO Molekulargewicht: 245,71 g/mol |
MDL Number:
MFCD00008284 CAS-Nummer: 1022-13-5 EINECS: 213-822-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...