Methyl-2-benzoylbenzoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL559830-25GEA 74.7 EUR
SIAL559830-25G
Methyl-2-benzoylbenzoat, Sigma-Aldrich®
Methyl-2-benzoylbenzoat
Formel: C₁₅H₁₂O₃
Molekulargewicht: 240,2540 g/mol
MDL Number: MFCD00017187
CAS-Nummer: 606-28-0
EINECS: 210-112-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR