Resodiacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1'-(4,6-Dihydroxy-1,3-phenylene)diethanone,4,6-Diacetylresorcinol,1,1'-(4,6-Dihydroxy-1,3-phenylene)bisethanone,4,6-Diacetyl-1,3-dihydroxybenzene

M-Clarity™ quality level = MQ100

391794-5G
SIAL391794-5GEA 71.9 EUR
SIAL391794-5G
Resodiacetophenon, Sigma-Aldrich®
Resodiacetophenon
Formel: C₁₀H₁₀O₄
Molekulargewicht: 194,1840 g/mol
MDL Number: MFCD00017655
CAS-Nummer: 2161-85-5
EINECS: 218-482-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR