Musk-Keton
Lieferant: EHRENSTORFER GMBH
EHERC15359000EA
189
EUR
EHERC15359000
Musk-Keton
Musk-Keton
Formel:
C₁₄H₁₈N₂O₅ |
MDL Number:
MFCD00024271 CAS-Nummer: 81-14-1 EINECS: 201-328-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...