4-Methoxybenzoesäureanhydrid

Lieferant: Thermo Scientific

CHMPBTB14618.250EA 0 EUR
CHMPBTB14618.250 MAYBBTB14618.1
4-Methoxybenzoesäureanhydrid
4-Methoxybenzoic anhydride
Formel: C₁₆H₁₄O₅
MDL Number: MFCD00017175
CAS-Nummer: 794-94-5
EINECS: 212-345-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR