5-Chlor-2-benzoxazolinon

Lieferant: Molekula

MOLE52084942-100GEA 0 EUR
MOLE52084942-100G
5-Chlor-2-benzoxazolinon
5-Chlor-2-benzoxazolinon
Formel: C₇H₄ClNO₂
Molekulargewicht: 169,57 g/mol
MDL Number: MFCD00005717
CAS-Nummer: 95-25-0
EINECS: 202-403-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR