6-Chlornicotinoylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL528102-25GEA 138 EUR
SIAL528102-25G
6-Chlornicotinoylchlorid, Sigma-Aldrich®
6-Chlornicotinoylchlorid
Formel: C₆H₃Cl₂NO
Molekulargewicht: 176,00 g/mol
MDL Number: MFCD00051775
CAS-Nummer: 58757-38-3
UN-Nummer: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR