2-Methoxyisonicotinamid

Lieferant: Thermo Scientific

MAYBSPB06368.1EA 0 EUR
MAYBSPB06368.1 MAYBSPB06368.10
2-Methoxyisonicotinamid
2-Methoxyisonicotinamid
Formel: C₇H₈N₂O₂
MDL Number: MFCD00174337
CAS-Nummer: 105612-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR