2,3,3-Trimethylindolenin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALT76805-25GEA 61.5 EUR
SIALT76805-25G
2,3,3-Trimethylindolenin, Sigma-Aldrich®
2,3,3-Trimethylindolenin
Formel: C₁₁H₁₃N
Molekulargewicht: 159,23
MDL Number: MFCD00005724
CAS-Nummer: 1640-39-7
EINECS: 216-685-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR