10,11-Dihydrocarbamazepin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL195421-100MGEA 66.4 EUR
SIAL195421-100MG
10,11-Dihydrocarbamazepin, Sigma-Aldrich®
10,11-Dihydrocarbamazepin
Formel: C₁₅H₁₄N₂O
Molekulargewicht: 238,29
MDL Number: MFCD00005072
CAS-Nummer: 3564-73-6
EINECS: 222-649-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR