2-Phenyl-2',4',6'-trihydroxyacetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL630632-1GEA
107
EUR
SIAL630632-1G
2-Phenyl-2',4',6'-trihydroxyacetophenon, Sigma-Aldrich®
2-Phenyl-2',4',6'-trihydroxyacetophenon
Formel:
C₁₄H₁₂O₄ Molekulargewicht: 244,2427 g/mol |
MDL Number:
MFCD00205539 CAS-Nummer: 727-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...