Dysprosium(III)acetat Tetrahydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL325538-25GEA 197 EUR
SIAL325538-25G
Dysprosium(III)acetat Tetrahydrat, Sigma-Aldrich®
Dysprosium(III)acetat Tetrahydrat
Formel: C₆H₉DyO₆·4H₂O
Molekulargewicht: 411,68 g/mol
MDL Number: MFCD00013033
CAS-Nummer: 15280-55-4
EINECS: 242-565-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR