Toluol-[Methyl-D3] (99% D), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ quality level = MQ200
SIAL487074-5GEA
798
EUR
SIAL487074-5G
Toluol-[Methyl-D3] (99% D), Sigma-Aldrich®
Toluol-[Methyl-D3]
Formel:
C₆H₅CD₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00084185 CAS-Nummer: 1124-18-1 UN-Nummer: 1294 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...