4'-tert-Butyl-2',6'-dimethylacetophenon, Sigma-Aldrich®

Lieferant: Merck

SIAL451495-1GEA 51.6 EUR
SIAL451495-1G
4'-tert-Butyl-2',6'-dimethylacetophenon, Sigma-Aldrich®
4'-tert-Butyl-2',6'-dimethylacetophenon
Formel: C₁₄H₂₀O
MDL Number: MFCD00026301
CAS-Nummer: 2040-10-0
EINECS: 218-037-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR