4-Amino-2,3,5,6-tetrafluorpyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL300624-1GEA
30.7
EUR
SIAL300624-1G
4-Amino-2,3,5,6-tetrafluorpyridin, Sigma-Aldrich®
4-Amino-2,3,5,6-tetrafluoropyridine
Formel:
C₅H₂F₄N₂ Molekulargewicht: 166,08 g/mol |
MDL Number:
MFCD00010855 CAS-Nummer: 1682-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...