3-Fluor-4-(trifluormethoxy)anilin 97%, JRD
|
Achtung
|
H32561.03EA
222
EUR
H32561.03
H32561.MD
3-Fluor-4-(trifluormethoxy)anilin 97%, JRD
3-Fluor-4-(trifluormethoxy)anilin
Formel:
C₇H₅F₄NO Molekulargewicht: 195,12 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832383 CAS-Nummer: 1017779-69-9 UN-Nummer: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...