Ethylpentafluorpropionat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL290920-25GEA 77.7 EUR
SIAL290920-25G
Ethylpentafluorpropionat, Sigma-Aldrich®
Ethylpentafluorpropionat
Formel: C₅H₅F₅O₂
Molekulargewicht: 192,09 g/mol
MDL Number: MFCD00000431
CAS-Nummer: 426-65-3
EINECS: 207-043-6
UN-Nummer: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR