2,2,2-Trifluorethylmethansulfonat, Sigma-Aldrich®

Lieferant: Merck

SIAL64297-5ML-FEA 73.6 EUR
SIAL64297-5ML-F
2,2,2-Trifluorethylmethansulfonat, Sigma-Aldrich®
2,2,2-Trifluorethylmethansulfonat
Formel: C₃H₅F₃O₃S
MDL Number: MFCD00025058
CAS-Nummer: 25236-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR