3',5'-Diethoxyacetophenon, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL574546-5GEA
30.6
EUR
SIAL574546-5G
3',5'-Diethoxyacetophenon, Sigma-Aldrich®
3',5'-Diethoxyacetophenon
Formel:
C₁₂H₁₆O₃ Molekulargewicht: 208,2536 g/mol |
MDL Number:
MFCD02258864 CAS-Nummer: 103604-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...