2-Ethinyltoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL558788-5GEA 348 EUR
SIAL558788-5G
2-Ethinyltoluol, Sigma-Aldrich®
2-Ethinyltoluol
Formel: C₉H₈
Molekulargewicht: 116,1598 g/mol
MDL Number: MFCD03427202
CAS-Nummer: 766-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR