Geranylacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ300

SIALW250902-10KG-KEA 499 EUR
SIALW250902-10KG-K
Geranylacetat, Sigma-Aldrich®
Geranylacetat
Formel: C₁₂H₂₀O₂
Molekulargewicht: 196,29 g/mol
MDL Number: MFCD00015037
CAS-Nummer: 105-87-3
EINECS: 203-341-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR