(3-Chlorpropyl)trimethylsilan, Sigma-Aldrich®

Lieferant: Merck

SIAL26253-5ML-FEA 162 EUR
SIAL26253-5ML-F
(3-Chlorpropyl)trimethylsilan, Sigma-Aldrich®
(3-Chloropropyl)trimethylsilane
Formel: C₆H₁₅ClSi
Molekulargewicht: 150,73 g/mol
MDL Number: MFCD00045299
CAS-Nummer: 2344-83-4
EINECS: 219-059-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR