2,4,6-T (2,4,6-Trichlorphenoxyessigsäure)

Lieferant: EHRENSTORFER GMBH

EHERC17777000EA 170 EUR
EHERC17777000
2,4,6-T (2,4,6-Trichlorphenoxyessigsäure)
2,4,6-T (2,4,6-Trichlorphenoxyessigsäure)
Formel: C₈H₅Cl₃O₃
MDL Number: MFCD00053498
CAS-Nummer: 575-89-3
EINECS: 209-394-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR